siRNA Details
Gene Symbol: Btbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011932.1 | NP_001011932.1 | 8 | 1465 | ||
GenBank mRNA | BC085850.1 | - | 8 | 1465 | 7189751 | |
FQ221559.1 | - | 4 | 473 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Btbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011932.1 | NP_001011932.1 | 8 | 1465 | ||
GenBank mRNA | BC085850.1 | - | 8 | 1465 | 7189751 | |
FQ221559.1 | - | 4 | 473 |