siRNA Details
| Gene Symbol: Fam154b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106277.1 | NP_001099747.1 | 2, 3 | 349 | ||
| XM_006229473.2 | XP_006229535.1 | 3, 4 | 560 | |||
| XM_006229474.2 | XP_006229536.1 | 3, 4 | 2252 | |||
| XM_006229475.2 | XP_006229537.1 | 4, 5 | 643 | |||
| XM_008759511.1 | XP_008757733.1 | 4, 5 | 1451 | |||

Fam154b
Genomic Map