siRNA Details
| Gene Symbol: Zfand6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007630.1 | NP_001007631.1 | 7 | 697 | ||
| XM_006229499.2 | XP_006229561.1 | 9 | 1002 | |||
| XM_006229500.2 | XP_006229562.1 | 7 | 682 | |||
| XM_006229502.2 | XP_006229564.1 | 8 | 825 | |||
| XM_006229504.1 | XP_006229566.1 | 8 | 836 | |||
| XM_006229505.2 | XP_006229567.1 | 7 | 690 | |||
| XM_006229506.2 | XP_006229568.1 | 7 | 687 | |||
| XM_006229507.2 | XP_006229569.1 | 6 | 648 | |||
| XM_006229508.2 | XP_006229570.1 | 6 | 643 | |||
| XM_008759611.1 | XP_008757833.1 | 7 | 2227 | |||
| GenBank mRNA | BC076394.1 | - | 7 | 697 | 7107784 | |

Zfand6
Genomic Map