siRNA Details
| Gene Symbol: Olr10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000115.1 | NP_001000115.1 | 1 | 184 | ||
| Gene Symbol: Olr11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000114.1 | NP_001000114.1 | 1 | 184 | ||

Olr10
Genomic Map