siRNA Details
| Gene Symbol: Olr19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000117.1 | NP_001000117.1 | 1 | 495 | ||
| Gene Symbol: LOC100910993 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003752171.3 | XP_003752219.1 | 1 | 564 | ||

Olr19
Genomic Map