siRNA Details
| Gene Symbol: Tmem135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013896.1 | NP_001013918.1 | 12 | 1219 | ||
| GenBank mRNA | BC085115.1 | - | 12 | 1219 | 7106476 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem135| Gene Symbol: Tmem135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013896.1 | NP_001013918.1 | 12 | 1219 | ||
| GenBank mRNA | BC085115.1 | - | 12 | 1219 | 7106476 | |