siRNA Details
| Gene Symbol: Ccdc89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134833.2 | NP_001128305.1 | 1 | 1048 | ||
| GenBank mRNA | BC167063.1 | - | 1 | 1048 | 7453194 | |
| Gene Symbol: LOC100911776 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003748922.3 | XP_003748970.1 | 1 | 2588 | ||

Ccdc89
Genomic Map