siRNA Details
| Gene Symbol: Tmem126b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106280.2 | NP_001099750.2 | 4, 5 | 512 | ||
| GenBank mRNA | BC167111.1 | - | 4, 5 | 512 | 6921139 | |
| FQ216464.1 | - | 4, 5 | 821 | |||
| FQ223509.1 | - | 4, 5 | 513 | |||

Tmem126b
Genomic Map