siRNA Details
| Gene Symbol: Nars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034921.1 | NP_001030093.1 | 3, 4 | 623 | ||
| GenBank mRNA | BC104691.1 | - | 3, 4 | 623 | 7107607 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nars2| Gene Symbol: Nars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034921.1 | NP_001030093.1 | 3, 4 | 623 | ||
| GenBank mRNA | BC104691.1 | - | 3, 4 | 623 | 7107607 | |