siRNA Details
| Gene Symbol: Nars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034921.1 | NP_001030093.1 | 6, 7 | 935 | ||
| GenBank mRNA | BC088466.1 | - | 92 | |||
| BC104691.1 | - | 6, 7 | 935 | 7107607 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nars2| Gene Symbol: Nars2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034921.1 | NP_001030093.1 | 6, 7 | 935 | ||
| GenBank mRNA | BC088466.1 | - | 92 | |||
| BC104691.1 | - | 6, 7 | 935 | 7107607 | ||