siRNA Details
| Gene Symbol: Ndufc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009290.1 | NP_001009290.1 | 2 | 340 | ||
| GenBank mRNA | BC086952.1 | - | 2 | 340 | 7325437 | |
| FQ220552.1 | - | 2 | 389 | |||
| FQ224080.1 | - | 2 | 385 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ndufc2| Gene Symbol: Ndufc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009290.1 | NP_001009290.1 | 2 | 340 | ||
| GenBank mRNA | BC086952.1 | - | 2 | 340 | 7325437 | |
| FQ220552.1 | - | 2 | 389 | |||
| FQ224080.1 | - | 2 | 385 | |||