siRNA Details
| Gene Symbol: Lrrc32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170434.1 | NP_001163905.1 | 3 | 1254 | ||
| XM_006229746.2 | XP_006229808.1 | 3 | 2104 | |||
| XM_006229747.2 | XP_006229809.1 | 3 | 2111 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc32| Gene Symbol: Lrrc32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170434.1 | NP_001163905.1 | 3 | 1254 | ||
| XM_006229746.2 | XP_006229808.1 | 3 | 2104 | |||
| XM_006229747.2 | XP_006229809.1 | 3 | 2111 | |||