siRNA Details
| Gene Symbol: Pold3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024750.1 | NP_001019921.1 | 2 | 182 | ||
| GenBank mRNA | BC098010.1 | - | 2 | 182 | 7454306 | |
| FQ211238.1 | - | 184 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pold3| Gene Symbol: Pold3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024750.1 | NP_001019921.1 | 2 | 182 | ||
| GenBank mRNA | BC098010.1 | - | 2 | 182 | 7454306 | |
| FQ211238.1 | - | 184 | ||||