siRNA Details
| Gene Symbol: Folr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106283.1 | NP_001099753.1 | 4 | 498 | ||
| XM_006229858.2 | XP_006229920.1 | 3 | 378 | |||
| XM_006229859.2 | XP_006229921.1 | 4 | 580 | |||
| XM_008759703.1 | XP_008757925.1 | 3 | 413 | |||
| GenBank mRNA | FQ211323.1 | - | 3 | 345 | ||

Folr2
Genomic Map