siRNA Details
| Gene Symbol: Far1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011933.1 | NP_001011933.1 | 4 | 554 | ||
| XM_006230017.2 | XP_006230079.1 | 5 | 636 | |||
| XM_006230018.2 | XP_006230080.1 | 5 | 561 | |||
| XM_006230020.2 | XP_006230082.1 | 5 | 638 | |||
| GenBank mRNA | BC082015.1 | - | 4 | 554 | 7132096 | |
| FQ221385.1 | - | 5 | 620 | |||

Far1
Genomic Map