siRNA Details
| Gene Symbol: Olr45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000124.1 | NP_001000124.1 | 1 | 832 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr45| Gene Symbol: Olr45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000124.1 | NP_001000124.1 | 1 | 832 | ||