siRNA Details
| Gene Symbol: Olr40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000127.1 | NP_001000127.1 | 1 | 529 | ||
| Gene Symbol: Olr39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000128.1 | NP_001000128.1 | 1 | 529 | ||

Olr40
Genomic Map