siRNA Details
| Gene Symbol: Olr62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000129.1 | NP_001000129.1 | 1 | 827 | ||
| Gene Symbol: Olr60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000748.1 | NP_001000748.1 | 1 | 827 | ||
| Gene Symbol: Olr61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001022.1 | NP_001001022.1 | 1 | 827 | ||

Olr62
Genomic Map