siRNA Details
| Gene Symbol: Olr68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000133.1 | NP_001000133.1 | 1 | 274 | ||
| Gene Symbol: Olr67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001279.1 | NP_001001279.1 | 1 | 244 | ||

Olr68
Genomic Map