siRNA Details
| Gene Symbol: Olr93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000141.1 | NP_001000141.1 | 1 | 518 | ||
| Gene Symbol: Olr95 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001024.1 | NP_001001024.1 | 1 | 518 | ||

Olr93
Genomic Map