siRNA Details
| Gene Symbol: Olr98 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000143.1 | NP_001000143.1 | 1 | 136 | ||
| Gene Symbol: Olr101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000744.1 | NP_001000744.1 | 1 | 139 | ||

Olr98
Genomic Map