siRNA Details
| Gene Symbol: Olr103 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000144.1 | NP_001000144.1 | 1 | 323 | ||
| Gene Symbol: Olr98 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000143.1 | NP_001000143.1 | 1 | 320 | ||

Olr103
Genomic Map