siRNA Details
| Gene Symbol: Olr126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000158.1 | NP_001000158.1 | 1 | 707 | ||
| Gene Symbol: Olr127 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001026.1 | NP_001001026.1 | 1 | 707 | ||

Olr126
Genomic Map