siRNA Details
Gene Symbol: Olr128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000159.1 | NP_001000159.1 | 1 | 273 |
Gene Symbol: LOC103694854 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773798.1 | XP_008772020.1 | 1 | 306 |
