siRNA Details
| Gene Symbol: Olr140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000162.1 | NP_001000162.1 | 1 | 593 | ||
| Gene Symbol: Olr137 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001283.1 | NP_001001283.1 | 1 | 611 | ||

Olr140
Genomic Map