siRNA Details
| Gene Symbol: USP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017415.1 | NP_001017415.1 | 7 | 1630 | ||
| NM_001017416.1 | NP_001017416.1 | 7 | 1535 | |||
| NM_003368.4 | NP_003359.3 | 7 | 2060 | |||
| GenBank mRNA | AB014458.1 | - | 7 | 1411 | ||
| AB208893.1 | - | 7 | 1691 | |||
| AF117386.1 | - | 7 | 1547 | |||
| AK312423.1 | - | 7 | 1723 | |||
| AL117575.1 | - | 6 | 1382 | |||
| BC023216.1 | - | 3 | 895 | |||
| BC050525.1 | - | 7 | 1764 | 6473568 | ||

USP1
Genomic Map