siRNA Details
| Gene Symbol: Ubqlnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013899.1 | NP_001013921.1 | 1 | 1364 | ||
| GenBank mRNA | BC083634.1 | - | 1 | 1364 | 7134647 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubqlnl| Gene Symbol: Ubqlnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013899.1 | NP_001013921.1 | 1 | 1364 | ||
| GenBank mRNA | BC083634.1 | - | 1 | 1364 | 7134647 | |