siRNA Details
| Gene Symbol: RGD1310717 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106287.1 | NP_001099757.1 | 6 | 1383 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1310717| Gene Symbol: RGD1310717 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106287.1 | NP_001099757.1 | 6 | 1383 | ||