siRNA Details
| Gene Symbol: Olr183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000181.1 | NP_001000181.1 | 1 | 349 | ||
| Gene Symbol: Olr179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000949.1 | NP_001000949.1 | 1 | 349 | ||
| Gene Symbol: Olr181 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001030.1 | NP_001001030.1 | 1 | 349 | ||

Olr183
Genomic Map