siRNA Details
| Gene Symbol: Olr189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000184.1 | NP_001000184.1 | 1 | 815 | ||
| Gene Symbol: Olr186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001031.1 | NP_001001031.1 | 1 | 815 | ||

Olr189
Genomic Map