siRNA Details
| Gene Symbol: Fam160a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005538.2 | NP_001005538.2 | 3 | 539 | ||
| XM_006229931.2 | XP_006229993.1 | 3 | 573 | |||
| XM_006229932.2 | XP_006229994.1 | 3 | 572 | |||
| XM_008759708.1 | XP_008757930.1 | 3 | 573 | |||
| GenBank mRNA | BC082011.1 | - | 3 | 525 | 7132039 | |

Fam160a2
Genomic Map