siRNA Details
| Gene Symbol: Fam160a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005538.2 | NP_001005538.2 | 2 | 428 | ||
| XM_006229931.2 | XP_006229993.1 | 2 | 462 | |||
| XM_006229932.2 | XP_006229994.1 | 2 | 461 | |||
| XM_008759708.1 | XP_008757930.1 | 2 | 462 | |||
| GenBank mRNA | BC082011.1 | - | 2 | 414 | 7132039 | |
| CV112456.1 | - | 2 | 428 | |||

Fam160a2
Genomic Map