siRNA Details
| Gene Symbol: Arfip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004222.1 | NP_001004222.1 | 6 | 775 | ||
| XM_006229933.2 | XP_006229995.1 | 6 | 793 | |||
| GenBank mRNA | BC079174.1 | - | 6 | 775 | 7130075 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arfip2| Gene Symbol: Arfip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004222.1 | NP_001004222.1 | 6 | 775 | ||
| XM_006229933.2 | XP_006229995.1 | 6 | 793 | |||
| GenBank mRNA | BC079174.1 | - | 6 | 775 | 7130075 | |