siRNA Details
| Gene Symbol: Olr206 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000193.1 | NP_001000193.1 | 1 | 407 | ||
| Gene Symbol: Olr204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000192.1 | NP_001000192.1 | 1 | 407 | ||

Olr206
Genomic Map