siRNA Details
| Gene Symbol: Olr231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000206.1 | NP_001000206.1 | 1 | 396 | ||
| XM_008759709.1 | XP_008757931.1 | 2 | 1105 | |||
| XM_008759710.1 | XP_008757932.1 | 3 | 1615 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr231| Gene Symbol: Olr231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000206.1 | NP_001000206.1 | 1 | 396 | ||
| XM_008759709.1 | XP_008757931.1 | 2 | 1105 | |||
| XM_008759710.1 | XP_008757932.1 | 3 | 1615 | |||