siRNA Details
| Gene Symbol: Olr235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000209.1 | NP_001000209.1 | 1 | 766 | ||
| Gene Symbol: Olr239 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000211.1 | NP_001000211.1 | 1 | 766 | ||

Olr235
Genomic Map