siRNA Details
| Gene Symbol: Olr240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229963.2 | XP_006230025.1 | 1 | 695 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olr240| Gene Symbol: Olr240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006229963.2 | XP_006230025.1 | 1 | 695 | ||