siRNA Details
Gene Symbol: VARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006295.2 | NP_006286.1 | 2 | 569 | ||
XM_005249362.2 | XP_005249419.1 | 2 | 562 | |||
GenBank mRNA | CX167000.1 | - | 1 | 85 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: VARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006295.2 | NP_006286.1 | 2 | 569 | ||
XM_005249362.2 | XP_005249419.1 | 2 | 562 | |||
GenBank mRNA | CX167000.1 | - | 1 | 85 |