siRNA Details
| Gene Symbol: Hbs1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011934.1 | NP_001011934.1 | 7 | 941 | ||
| GenBank mRNA | BC079463.1 | - | 7 | 941 | 7122195 | |
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_019702, NM_001042593, AK168671, BC010251, AK166140, and others.

Hbs1l
Genomic Map