siRNA Details
| Gene Symbol: Hbs1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011934.1 | NP_001011934.1 | 3 | 282 | ||
| GenBank mRNA | BC079463.1 | - | 3 | 282 | 7122195 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hbs1l| Gene Symbol: Hbs1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011934.1 | NP_001011934.1 | 3 | 282 | ||
| GenBank mRNA | BC079463.1 | - | 3 | 282 | 7122195 | |