siRNA Details
| Gene Symbol: Tmem9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106289.1 | NP_001099759.1 | 2, 3 | 371 | ||
| XM_006229972.2 | XP_006230034.1 | 2, 3 | 1662 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_020050, BC002208, AK009147, AK077253, AK147127, and others.

Tmem9b
Genomic Map