siRNA Details
| Gene Symbol: Tmem9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106289.1 | NP_001099759.1 | 4 | 536 | ||
| XM_006229972.2 | XP_006230034.1 | 4 | 1827 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem9b| Gene Symbol: Tmem9b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106289.1 | NP_001099759.1 | 4 | 536 | ||
| XM_006229972.2 | XP_006230034.1 | 4 | 1827 | |||