siRNA Details
| Gene Symbol: Eif3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277302.1 | NP_001264231.1 | 5, 6 | 787 | ||
| GenBank mRNA | FQ212642.1 | - | 5, 6 | 776 | ||
| FQ213714.1 | - | 5, 6 | 788 | |||
| FQ229379.1 | - | 5, 6 | 461 | |||
| FQ230130.1 | - | 5, 6 | 775 | |||

Eif3f
Genomic Map