siRNA Details
| Gene Symbol: Palb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191603.1 | NP_001178532.1 | 6 | 2346 | ||
| XM_008759711.1 | XP_008757933.1 | 4 | 3522 | |||
| XM_008759713.1 | XP_008757935.1 | 6 | 2502 | |||
| GenBank mRNA | FQ211178.1 | - | 6 | 2460 | ||

Palb2
Genomic Map