siRNA Details
| Gene Symbol: Cog7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033889.1 | NP_001029061.1 | 4, 5 | 706 | ||
| GenBank mRNA | BC101931.1 | - | 4, 5 | 706 | 7938773 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cog7| Gene Symbol: Cog7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033889.1 | NP_001029061.1 | 4, 5 | 706 | ||
| GenBank mRNA | BC101931.1 | - | 4, 5 | 706 | 7938773 | |