siRNA Details
| Gene Symbol: Ypel3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135698.1 | NP_001129170.1 | 4, 5 | 465 | ||
| XM_006230233.1 | XP_006230295.1 | 4, 5 | 742 | |||
| XM_006230235.1 | XP_006230297.1 | 4, 5 | 431 | |||
| GenBank mRNA | CB582520.1 | - | 4, 5 | 477 | ||
| DV728986.1 | - | 1, 2 | 82 | |||
| FQ229460.1 | - | 4, 5 | 454 | |||
| FQ229697.1 | - | 4, 5 | 454 | |||

Ypel3
Genomic Map