siRNA Details
| Gene Symbol: Fam57b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106296.2 | NP_001099766.2 | 4 | 626 | ||
| XM_006230237.2 | XP_006230299.1 | 5 | 702 | |||
| XM_006230239.2 | XP_006230301.1 | 4 | 447 | |||
| XM_006230240.2 | XP_006230302.1 | 4 | 1222 | |||
| GenBank mRNA | BC161920.1 | - | 4 | 626 | 8370175 | |

Fam57b
Genomic Map