siRNA Details
| Gene Symbol: Ino80e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013900.1 | NP_001013922.1 | 7 | 745 | ||
| XM_006230243.1 | XP_006230305.1 | 7 | 668 | |||
| XM_006230244.1 | XP_006230306.1 | 7 | 620 | |||
| XM_006230245.1 | XP_006230307.1 | 6 | 554 | |||
| GenBank mRNA | BC079045.1 | - | 7 | 745 | 7115071 | |

Ino80e
Genomic Map