siRNA Details
Gene Symbol: LOC102553866 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006230369.2 | XP_006230431.1 | 3 | 1222 |
Gene Symbol: LOC102553962 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006230370.2 | XP_006230432.1 | 3 | 1101 |
