siRNA Details
| Gene Symbol: Pdilt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013902.1 | NP_001013924.1 | 8 | 1268 | ||
| GenBank mRNA | BC083897.1 | - | 8 | 1268 | 7133354 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdilt| Gene Symbol: Pdilt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013902.1 | NP_001013924.1 | 8 | 1268 | ||
| GenBank mRNA | BC083897.1 | - | 8 | 1268 | 7133354 | |